4-Phenylbutyronitrile 97%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
L01638.06
L01638.14
L01638.06EA
285
DKK
L01638.06
L01638.14
4-Phenylbutyronitrile 97%
4-Phenylbutyronitrile
Purity: 97%
CAS No.: 2046-18-6
Molecular Formula: C6H5(CH2)3CN
Formula Weight: 145.20
Flash Point: 105°(221°F)
Refractive Index: 1.5143
Boiling Point: 129-131°/10mm
Beilstein Reference: 2043002
UN No.: UN3276
MDL No.: MFCD00001973
Formel:
C₁₀H₁₁N Molekylvægt: 145.21 g/mol Kogepunkt: 129…131 °C (10 mmHg) Massefylde: 0.976 Flammepunkt: 105 °C (221 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00001973 CAS-nummer: 2046-18-6 EINECS: 218-068-7 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...