Trimethylsulphonium tetrafluoroborate ≥98%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
L00647.06
L00647.14
L00647.06EA
2600
DKK
L00647.06
L00647.14
Trimethylsulphonium tetrafluoroborate ≥98%
Trimethylsulphonium tetrafluoroborate
Purity: 98+%
CAS No.: 676-88-0
Molecular Formula: (CH3)3SBF4
Formula Weight: 163.98
Storage Temperature: 2°C to 8°C
Melting Point: 214-218°
Beilstein Reference: 3708437
UN No.: UN3261
MDL No.: MFCD00031582
Formel:
(CH₃)₃SBF₄ Smeltepunkt: 214…218 °C |
MDL Number:
MFCD00031582 CAS-nummer: 676-88-0 EINECS: 211-632-3 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...