Guld supplied in 0.1 mg/ml sodium citrate with stabilizer stabilised, nanoparticles 50 nm OD1 (Abs. 535 nm)
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
J67433.AC J67433.AE
J67433.ACEA 1190 DKK
J67433.AC J67433.AE
Guld supplied in 0.1 mg/ml sodium citrate with stabilizer stabilised, nanoparticles 50 nm OD1 (Abs. 535 nm)
Guld
CAS No.: 7440-57-5 Storage Temperature: 2°C to 8°C Physical Form: Liquid Appearance: Pink to red MDL No.: MFCD00003436
Applications: For adsorption of thiolated ligands such as oligonucleotides and polyethylene glycols. Also, due to the increased stability, these gold nanoparticles are ideal for use in physical and optical applications.
Formel: Au
Molekylvægt: 196.97 g/mol
Kogepunkt: 2800
Smeltepunkt: 1063
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00003436
CAS-nummer: 7440-57-5
Merck Index: 12,04526