Guld 99% supplied in 0.1 mM PBS, nanoparticles 50 nm reactant-free OD1 (Abs. 535 nm)
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
J67432.AE J67432.AC
J67432.AEEA 4120 DKK
J67432.AE J67432.AC
Guld 99% supplied in 0.1 mM PBS, nanoparticles 50 nm reactant-free OD1 (Abs. 535 nm)
Guld
Purity: 99% CAS No.: 7440-57-5 Storage Temperature: 2°C to 8°C Physical Form: Liquid Appearance: Red to pink MDL No.: MFCD00003436
Formel: Au
Molekylvægt: 196.97 g/mol
Kogepunkt: 2800
Smeltepunkt: 1063
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00003436
CAS-nummer: 7440-57-5
Merck Index: 12,04526