Guld 95% supplied in 0.1 mM PBS, nanoparticles 40 nm OD1 (Abs. 530 nm)
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
J67409.AC J67409.AE
J67409.ACEA 1240 DKK
J67409.AC J67409.AE
Guld 95% supplied in 0.1 mM PBS, nanoparticles 40 nm OD1 (Abs. 530 nm)
Guld
Purity: 95% CAS No.: 7440-57-5 Storage Temperature: 2°C to 8°C Physical Form: Liquid Appearance: Red to pink MDL No.: MFCD00003436
Applications: For development of peptide and protein gold conjugates for use in applications such as blotting, microscopy and TEM
Formel: Au
Molekylvægt: 196.97 g/mol
Kogepunkt: 2800
Smeltepunkt: 1063
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00003436
CAS-nummer: 7440-57-5
Merck Index: 12,04526