Guld supplied in 0.1 mg/ml sodium citrate with stabilizer stabilised, nanoparticles 40 nm OD1 (Abs. 530 nm)
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
J67001.AC J67001.AE
J67001.ACEA 1220 DKK
J67001.AC J67001.AE
Guld supplied in 0.1 mg/ml sodium citrate with stabilizer stabilised, nanoparticles 40 nm OD1 (Abs. 530 nm)
Guld
CAS No.: 7440-57-5 Storage Temperature: 2°C to 8°C Physical Form: Liquid Appearance: Red to pink Odour: Odourless MDL No.: MFCD00003436
Formel: Au
Molekylvægt: 196.97 g/mol
Kogepunkt: 2800
Smeltepunkt: 1063
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00003436
CAS-nummer: 7440-57-5
Merck Index: 12,04526