Cromolyn sodium 98%
Leverandør: Thermo Fisher ScientificSamlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
5,5'-[(2-Hydroxy-1,3-propanediyl)bis(oxy)]bis(4-oxo-4H-1-benzopyran-2-carboxylic Acid) Disodium Salt, Sodium 5,5'-((2-hydroxypropane-1,3-diyl)bis(oxy))bis(4-oxo-4H-chromene-2-carboxylate), Disodium Cromoglycate
Purity: 98%
CAS No.: 15826-37-6
Molecular Formula: C23H14Na2O11
Formula Weight: 512.33
Physical Form: Powder
Appearance: White to pale yellow
Solubility: Soluble in water to 100mg/ml
MDL No.: MFCD00057744
Applications: Mast cell stabilizer that prevents release of histamine from mast cells
Formel:
C₂₃H₁₆O₁₁ Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00057744 CAS-nummer: 15826-37-6 EINECS: 239-926-7 Merck Index: 14,2590 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...