Urapidil hydrochloride ≥98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

J63219.ME J63219.03
J63219.MEEA 1660 DKK
J63219.ME J63219.03
Urapidil hydrochloride ≥98%
Urapidil hydrochloride

Purity: 98+% CAS No.: 64887-14-5 Molecular Formula: C20H29N5O3.HCl Formula Weight: 423.93 Physical Form: Powder Appearance: White MDL No.: MFCD00078601


Applications: alpha-1-Adrenoceptor antagonist and 5-HT1A receptor agonist

Formel: C₂₀H₂₉N₅O₃·HCl
Molekylvægt: 423.93 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00078601
CAS-nummer: 64887-14-5

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR