PIPES-buffered saline (5X), pH 6,5

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

J63152.AK J63152.AP
J63152.AKEA 684 DKK
J63152.AK J63152.AP
PIPES-buffered saline (5X), pH 6,5
Buffere Biologiske buffere

CAS No.: 5625-37-6
Physical Form: Liquid
Appearance: Colourless
MDL No.: MFCD00006159

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR