Vasopressin ≥98%
Leverandør: Thermo Fisher ScientificSamlet vurdering
Gennemsntilig vurdering
Purity: 98+%
CAS No.: 11000-17-2
Molecular Formula: C46H65N15O12S2
Formula Weight: 1084.24
Storage Temperature: -30°C to -10°C
Physical Form: Lyophilized powder
Appearance: White
MDL No.: MFCD00076738
Applications: Has antidiuretic activity and plays a role in water balance in animals
Formel:
C₄₆H₆₅N₁₅O₁₂S₂ |
MDL Number:
MFCD00076738 CAS-nummer: 11000-17-2 EINECS: 234-236-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...