Miconazole nitrate ≥98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 1-[2-(2,4-Dichlorobenzyloxy)-2-(2,4-dichlorophenyl)ethyl]-1H-imidazole Nitrate

J60796.06 J60796.14
J60796.06EA 1620 DKK
J60796.06 J60796.14
Miconazole nitrate ≥98%
Miconazole nitrate

Purity: 98+% CAS No.: 22832-87-7 Molecular Formula: C18H14Cl4N2O.HNO3 Formula Weight: 479.15 Physical Form: Powder Appearance: White to off-white Melting Point: 170-185° Solubility: Slightly soluble in water. Soluble in propylene glycol or pyridine. MDL No.: MFCD00058161


Applications: Inhibits cytochrome P450-dependent 14a-demethylase

Formel: C₁₈H₁₄Cl₄N₂O·HNO₃
Molekylvægt: 479.14 g/mol
Smeltepunkt: 170…185 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00058161
CAS-nummer: 22832-87-7
EINECS: 245-256-6
Merck Index: 14,6178

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR