Loratadine ≥98%
Leverandør: Thermo Fisher ScientificSamlet vurdering
Gennemsntilig vurdering
Synonymer:
Ethyl 4-(8-chloro-5,6-dihydro-11H-benzo[5,6]cyclohepta[1,2-b]pyridin-11-ylidene)piperidine-1-carboxylate
Purity: 98+%
CAS No.: 79794-75-5
Molecular Formula: C22H23ClN2O2
Formula Weight: 382.88
Physical Form: Powder
Appearance: White
Melting Point: 134°
MDL No.: MFCD00672869
Applications: A nonsedating-type histamine H1-receptor antagonist
Formel:
C₂₂H₂₃ClN₂O₂ Smeltepunkt: 134 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00672869 CAS-nummer: 79794-75-5 Merck Index: 14,5578 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...