Lactulose 99%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
J60160.14
J60160.18
J60160.22
J60160.14EA
2020
DKK
J60160.14
J60160.18
J60160.22
Lactulose 99%
Lactulose
Purity: 99%
CAS No.: 4618-18-2
Molecular Formula: C12H22O11
Formula Weight: 324.30
Physical Form: Powder
Density: 1.32
Appearance: White
Melting Point: 169°
Beilstein Reference: 93773
MDL No.: MFCD00151469
Applications: A synthetic sugar
Formel:
C₁₂H₂₂O₁₁ Smeltepunkt: 169 °C Massefylde: 1,32 Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00151469 CAS-nummer: 4618-18-2 EINECS: 225-027-7 Merck Index: 14,5346 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...