1-Methylguanidine xhydrochloride 98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Methylguanidine hydrochloride

J60033.06 J60033.14
J60033.06EA 859 DKK
J60033.06 J60033.14
1-Methylguanidine xhydrochloride 98%
1-Methylguanidine xhydrochloride

Purity: 98% CAS No.: 22661-87-6 Molecular Formula: C2H7N3.HCl Formula Weight: 109.56 Storage Temperature: -30°C to -10°C Physical Form: Crystalline powder Sensitivity: Hygroscopic Appearance: White to off-white MDL No.: MFCD00012576

Formel: C₂H₈ClN₃
MDL Number: MFCD00012576
CAS-nummer: 22661-87-6
EINECS: 245-147-3

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR