Brooker's merocyanine indicator dyed

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

H66517.MD H66517.03
H66517.MDEA 2610 DKK
H66517.MD H66517.03
Brooker's merocyanine indicator dyed
Brooker's merocyanine

Brooker's merocyanine dye is used as solvent polarity indicators. It is used as pH sensors and transition metal cation indicators.

Formel: C₁₄H₁₃NO
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00010099
CAS-nummer: 23302-83-2

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR