2-Brom-5-hydroxybenzaldehyd 95%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

H64884.06 H64884.14 H64884.22
H64884.06EA 642 DKK
H64884.06 H64884.14 H64884.22
2-Brom-5-hydroxybenzaldehyd 95%
2-Bromo-5-hydroxybenzaldehyde

Purity: 95% CAS No.: 2973-80-0 Molecular Formula: C7H5BrO2 Formula Weight: 201.02 Physical Form: Solid Melting Point: 130-135° Beilstein Reference: 2079921 UN No.: UN3077 MDL No.: MFCD03033065

Formel: C₇H₅BrO₂
Smeltepunkt: 130…135 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD03033065
CAS-nummer: 2973-80-0
ADR: 9,III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR