Methyl2-chlornicotinat 98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Methyl 2-chloronicotinate, Methyl 2-chloropyridine-3-carboxylate

H64798.06 H64798.14 H64798.22
H64798.06EA 473 DKK
H64798.06 H64798.14 H64798.22
Methyl2-chlornicotinat 98%
Methyl2-chlornicotinat

Purity: 98% CAS No.: 40134-18-7 Molecular Formula: C7H6ClNO2 Formula Weight: 171.58 Flash Point: 110°(230°F) Physical Form: Liquid Density: 1.314 Refractive Index: 1.5370 Appearance: Colourless Boiling Point: 150°/1mm Beilstein Reference: 122129 MDL No.: MFCD01318629

Formel: C₇H₆ClNO₂
Molekylvægt: 171.58 g/mol
Kogepunkt: 150 °C (1 mmHg)
Massefylde: 1.314
Flammepunkt: >110 °C (230 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD01318629
CAS-nummer: 40134-18-7
EINECS: 254-805-9

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR