1-Cyclopentylethanone 98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

H64744.03 H64744.06
H64744.03EA 1180 DKK
H64744.03 H64744.06
1-Cyclopentylethanone 98%
1-Cyclopentylethanone

Purity: 98% CAS No.: 6004-60-0 Molecular Formula: C7H12O Formula Weight: 112.17 Physical Form: Liquid Density: 0.913 Refractive Index: 1.4435 Appearance: Colourless Boiling Point: 151-156° Beilstein Reference: 1853483 MDL No.: MFCD00060799

Formel: C₇H₁₂O
Molekylvægt: 112.172 g/mol
Kogepunkt: 151…156 °C
Massefylde: 0.913
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00060799
CAS-nummer: 6004-60-0

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR