4-Chloro-2-fluoropyridine 95%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
H64557.03
H64557.06
H64557.03EA
2680
DKK
H64557.03
H64557.06
4-Chloro-2-fluoropyridine 95%
4-Chloro-2-fluoropyridine
Purity: 95%
CAS No.: 34941-92-9
Molecular Formula: C5H3ClFN
Formula Weight: 131.53
Physical Form: Solid
Beilstein Reference: 1634727
MDL No.: MFCD04112503
Formel:
C₅H₃ClFN Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD04112503 CAS-nummer: 34941-92-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...