5-formylthiophene-2-carbonitrile 95%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 5-Cyanothiophene-2-carboxaldehyde

H64462.03 H64462.06
H64462.03EA 862 DKK
H64462.03 H64462.06
5-formylthiophene-2-carbonitrile 95%
5-Formylthiophene-2-carbonitrile

Purity: 95% CAS No.: 21512-16-3 Molecular Formula: C6H3NOS Formula Weight: 137.16 Physical Form: Solid Appearance: White Melting Point: 90-95° Beilstein Reference: 115517 UN No.: UN3439 MDL No.: MFCD09037804

Formel: C₆H₃NOS
Molekylvægt: 137.156 g/mol
Smeltepunkt: 90…95 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD09037804
CAS-nummer: 21512-16-3
ADR: 6.1 III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR