6-Azaindol ≥97%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
H64458.03
H64458.06
H64458.03EA
1140
DKK
H64458.03
H64458.06
6-Azaindol ≥97%
6-Azaindol
Purity: 98+%
CAS No.: 271-29-4
Molecular Formula: C7H6N2
Formula Weight: 118.14
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Sensitivity: Air and Light Sensitive
Beilstein Reference: 109685
MDL No.: MFCD01686899
Formel:
C₇H₆N₂ Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD01686899 CAS-nummer: 271-29-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...