1-Pyreneboronic acid 95%
Leverandør: Thermo Fisher ScientificSamlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Purity: 95%
CAS No.: 164461-18-1
Molecular Formula: C16H11BO2
Formula Weight: 246.07
Physical Form: Powder
Appearance: Pale yellow
Melting Point: 247-251° lit.
Beilstein Reference: 7134073
MDL No.: MFCD04974062
Formel:
C₁₆H₁₁BO₂ Smeltepunkt: 247…251 °C (lit.) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD04974062 CAS-nummer: 164461-18-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...