2-Chloro-1-nitro-3-(trifluoromethyl)benzene 98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2-Chloro-3-nitrobenzotrifluoride

H64051.03 H64051.06
H64051.03EA 698 DKK
H64051.03 H64051.06
2-Chloro-1-nitro-3-(trifluoromethyl)benzene 98%
2-Chloro-1-nitro-3-(trifluoromethyl)benzene

Purity: 98% CAS No.: 39974-35-1 Molecular Formula: C7H3ClF3NO2 Formula Weight: 225.55 Flash Point: 110°(230°F) Physical Form: Solid Melting Point: 25-30° Beilstein Reference: 10429463 MDL No.: MFCD14525509

Formel: C₇H₃ClF₃NO₂
Molekylvægt: 225.551 g/mol
Smeltepunkt: 25…30 °C
Flammepunkt: >110 °C (230 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD14525509
CAS-nummer: 39974-35-1
EINECS: 254-724-9

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR