2-Brom-m-toluidin 97+%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

H64014.MD H64014.03
H64014.MDEA 507 DKK
H64014.MD H64014.03
2-Brom-m-toluidin 97+%
2-Brom-m-toluidin

Purity: 97+% CAS No.: 54879-20-8 Molecular Formula: C7H8BrN Formula Weight: 186.05 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Physical Form: Liquid Density: 1.489 Refractive Index: 1.6050 Appearance: Pale brown Boiling Point: 48°/0.03mm Beilstein Reference: 2079474 UN No.: UN2810 MDL No.: MFCD06411367

Formel: C₇H₈BrN
Molekylvægt: 186.05 g/mol
Kogepunkt: 48 °C (0.03 mmHg)
Massefylde: 1.489
Flammepunkt: >110 °C (230 °F)
MDL Number: MFCD06411367
CAS-nummer: 54879-20-8
ADR: 6.1 III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR