1-Boc-indoline-7-carboxylic acid 97%
Leverandør: Thermo Fisher ScientificSamlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Purity: 97%
CAS No.: 143262-20-8
Molecular Formula: C14H17NO4
Formula Weight: 263.29
Physical Form: Solid
Sensitivity: Light Sensitive
Appearance: Pale Yellow
Melting Point: 182° dec.
UN No.: UN3077
MDL No.: MFCD04973983
Formel:
C₁₄H₁₇NO₄ Smeltepunkt: 182 °C (dec.) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD04973983 CAS-nummer: 143262-20-8 ADR: 9,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...