H-Aib-OtBu·HCl 98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: tert-Butyl 2-aminoisobutyrate hydrochloride, tert-Butyl 2-amino-2-methylpropanoate hydrochloride

H63817.03 H63817.06
H63817.03EA 2070 DKK
H63817.03 H63817.06
H-Aib-OtBu·HCl 98%
H-Aib-OtBu·HCl

Purity: 98% CAS No.: 4512-32-7 Molecular Formula: C8H17NO2.HCl Formula Weight: 195.69 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White MDL No.: MFCD08272284

Formel: C₈H₁₇NO₂
Molekylvægt: 195.7 g/mol
MDL Number: MFCD08272284
CAS-nummer: 4512-32-7

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR