(2R,2'R)-Dimethyl-3,3'-disulphanediylbis(2-aminopropanoate) dihydrochloride 98%
Leverandør: Thermo Fisher ScientificSamlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Purity: 98%
CAS No.: 32854-09-4
Molecular Formula: C8H16N2O4S2.2HCl
Formula Weight: 341.28
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White
Melting Point: 182-183° dec
Beilstein Reference: 4727524
Optical Rotation: -36°(c=4 in methanol)
MDL No.: MFCD00012490
Formel:
C₈H₁₈Cl₂N₂O₄S₂ Smeltepunkt: 182…183 °C (dec.) Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD00012490 CAS-nummer: 32854-09-4 EINECS: 251-261-4 |