2-Iodpyridin 95%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
H63062.06
H63062.14
H63062.06EA
1160
DKK
H63062.06
H63062.14
2-Iodpyridin 95%
2-Iodopyridine
Purity: 95%
CAS No.: 5029-67-4
Molecular Formula: C5H4IN
Formula Weight: 205
Physical Form: Solid
Density: 1.928
Refractive Index: 1.632
Sensitivity: Light Sensitive
Appearance: Pale yellow
Melting Point: 52° dec
Beilstein Reference: 105790
MDL No.: MFCD00464928
Formel:
C₅H₄IN Smeltepunkt: 52 °C (dec.) Massefylde: 1,928 Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00464928 CAS-nummer: 5029-67-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...