3,4-Dimethoxythiophene 98%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
H56674.03
H56674.06
H56674.03EA
792
DKK
H56674.03
H56674.06
3,4-Dimethoxythiophene 98%
3,4-Dimethoxythiophene
Purity: 98%
CAS No.: 51792-34-8
Molecular Formula: C6H8O2S
Formula Weight: 144.19
Flash Point: 106°(223°F)
Refractive Index: 1.5409
Boiling Point: 100-102°/10-11mm
MDL No.: MFCD01096546
Formel:
C6H8O2S Kogepunkt: 100…102 °C (10…11 mmHg) Massefylde: 1,209 Flammepunkt: 106 °C (223 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD01096546 CAS-nummer: 51792-34-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...