1,10-Phenanthroline-5,6-dione 98%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
H55747.MD
H55747.03
H55747.06
H55747.MDEA
871
DKK
H55747.MD
H55747.03
H55747.06
1,10-Phenanthroline-5,6-dione 98%
1,10-Phenanthroline-5,6-dione
Purity: 98%
CAS No.: 27318-90-7
Molecular Formula: C12H6N2O2
Formula Weight: 210.19
Melting Point: 260° dec.
MDL No.: MFCD00014473
Formel:
C₁₂H₆N₂O₂ Smeltepunkt: 260 °C (dec.) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00014473 CAS-nummer: 27318-90-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...