4,6-Dimethyldibenzothiophene 97%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

H55472.MD H55472.03 H55472.06
H55472.MDEA 633 DKK
H55472.MD H55472.03 H55472.06
4,6-Dimethyldibenzothiophene 97%
4,6-Dimethyldibenzothiophene

Purity: 97% CAS No.: 1207-12-1 Molecular Formula: C14H12S Formula Weight: 212.31 Melting Point: 153-157° MDL No.: MFCD00216264

Formel: C₁₄H₁₂S
Smeltepunkt: 153…157 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00216264
CAS-nummer: 1207-12-1
EINECS: 214-894-7

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR