1-Ethynylcyclohexylamine 98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

H53514.06 H53514.14
H53514.06EA 1530 DKK
H53514.06 H53514.14
1-Ethynylcyclohexylamine 98%
1-Ethynylcyclohexylamine

Purity: 98% CAS No.: 30389-18-5 Molecular Formula: C8H13N Formula Weight: 123.20 Flash Point: 42°(108°F) Refractive Index: 1.4820 Boiling Point: 65-66°/20mm UN No.: UN2734 MDL No.: MFCD00001489

Formel: C₈H₁₃N
Kogepunkt: 65…66 °C (20 mmHg)
Massefylde: 0,913
Flammepunkt: 42 °C (108 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00001489
CAS-nummer: 30389-18-5
EINECS: 250-172-8
ADR: 8,II

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR