(2-Bromophenylethynyl)trimethylsilane 98%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
((2-Bromophenyl)ethynyl)trimethylsilane
H53402.03
H53402.06
H53402.14
H53402.03EA
424
DKK
H53402.03
H53402.06
H53402.14
(2-Bromophenylethynyl)trimethylsilane 98%
(2-Bromophenylethynyl)trimethylsilane
Purity: 98%
CAS No.: 38274-16-7
Molecular Formula: C11H13BrSi
Formula Weight: 253.21
Flash Point:
Formel:
C₁₁H₁₃BrSi Kogepunkt: 60…80 °C (2 mmHg) Massefylde: 1,19 Flammepunkt: >113 °C (235 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD01321391 CAS-nummer: 38274-16-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...