Boc-L-3,3-diphenylalanine 95%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
(S)-2-((tert-Butoxycarbonyl)amino)-3,3-diphenylpropanoic acid
H52063.06
H52063.03
H52063.06EA
4810
DKK
H52063.06
H52063.03
Boc-L-3,3-diphenylalanine 95%
Boc-L-3,3-diphenylalanine
Purity: 95%
CAS No.: 138662-63-2
Molecular Formula: C20H23NO4
Formula Weight: 341.41
Melting Point: 160° dec.
Beilstein Reference: 4880641
MDL No.: MFCD00191186
Formel:
C₂₀H₂₃NO₄ Smeltepunkt: 160 °C (dec.) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00191186 CAS-nummer: 138662-63-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...