Fmoc-3-(3-pyridyl)-L-alanine 95%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
Fmoc-3-Pal-OH, Fmoc-β-(3-pyridyl)-Ala-OH
H52056.03
H52056.06
H52056.03EA
2590
DKK
H52056.03
H52056.06
Fmoc-3-(3-pyridyl)-L-alanine 95%
Fmoc-3-(3-pyridyl)-L-alanine
Purity: 95%
CAS No.: 175453-07-3
Molecular Formula: C23H20N2O4
Formula Weight: 388.42
Beilstein Reference: 9154190
MDL No.: MFCD00144887
Formel:
C₂₃H₂₀N₂O₄ Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00144887 CAS-nummer: 175453-07-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...