Fmoc-L-4,4'-biphenylalanine 95%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Fmoc-4-phenyl-Phe-OH, Fmoc-Bip(4,4')-OH

H52002.MD H52002.03 H52002.06
H52002.MDEA 956 DKK
H52002.MD H52002.03 H52002.06
Fmoc-L-4,4'-biphenylalanine 95%
Fmoc-L-4,4'-biphenylalanine

Purity: 95% CAS No.: 199110-64-0 Molecular Formula: C30H25NO4 Formula Weight: 463.52 MDL No.: MFCD00191198

Formel: C₃₀H₂₅NO₄
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00191198
CAS-nummer: 199110-64-0

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR