4-(Methoxymethyl)piperidine hydrochloride

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

H51020.MD H51020.03
H51020.MDEA 1250 DKK
H51020.MD H51020.03
4-(Methoxymethyl)piperidine hydrochloride
4-(Methoxymethyl)piperidine hydrochloride

CAS No.: 916317-00-5 Molecular Formula: C7H15NO.HCl Formula Weight: 165.66 MDL No.: MFCD08062602

Formel: C₇H₁₆ClNO
Molekylvægt: 165.66 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD08062602
CAS-nummer: 916317-00-5

Bestil nu

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR