4-(Dimethylamino)butylamine 98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

H50276.03 H50276.06
H50276.03EA 1150 DKK
H50276.03 H50276.06
4-(Dimethylamino)butylamine 98%
4-(Dimethylamino)butylamine

Purity: 98% CAS No.: 3529-10-0 Molecular Formula: C6H16N2 Formula Weight: 116.21 Refractive Index: 1.4400 UN No.: UN2735 MDL No.: MFCD00047977

Formel: C₆H₁₆N₂
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00047977
CAS-nummer: 3529-10-0
EINECS: 222-559-1
ADR: 8,III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR