3-Acetylquinoline 97%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

H33206.03 H33206.06
H33206.03EA 946 DKK
H33206.03 H33206.06
3-Acetylquinoline 97%
3-Acetylquinoline

Purity: 97% CAS No.: 33021-53-3 Molecular Formula: C11H9NO Formula Weight: 171.20 Melting Point: 97-99° Boiling Point: 139°/4mm MDL No.: MFCD00030698

Formel: C₁₁H₉NO
Molekylvægt: 171.20 g/mol
Kogepunkt: 139 °C (4 mmHg)
Smeltepunkt: 97…99 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00030698
CAS-nummer: 33021-53-3

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR