(E)-5-(2-Bromovinyl)-2'-deoxyuridine 98%
Leverandør: Thermo Fisher ScientificSamlet vurdering
Gennemsntilig vurdering
Purity: 98%
CAS No.: 69304-47-8
Molecular Formula: C11H13BrN2O5
Formula Weight: 333.14
Storage Temperature: 2°C to 8°C
Melting Point: ca 165° dec.
Optical Rotation: +10° (c=1 in methanol)
MDL No.: MFCD00058585
Formel:
C₁₁H₁₃BrN₂O₅ Smeltepunkt: ∼ 165 °C (dec.) |
MDL Number:
MFCD00058585 CAS-nummer: 69304-47-8 |