4-Cyanopiperidine 96%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
H27722.14
H27722.06
H27722.14EA
2630
DKK
H27722.14
H27722.06
4-Cyanopiperidine 96%
4-Cyanopiperidine
Purity: 96%
CAS No.: 4395-98-6
Molecular Formula: C6H10N2
Formula Weight: 110.16
Flash Point: 91°(196°F)
Refractive Index: 1.4755
Odour: Amine-like
Boiling Point: 226°
UN No.: UN3276
MDL No.: MFCD05022468
Formel:
C₆H₁₀N₂ Kogepunkt: 226 °C Massefylde: 0,98 Flammepunkt: 91 °C (196 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD05022468 CAS-nummer: 4395-98-6 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...