3-(Trifluoromethyl)piperidine 97%
Leverandør: Thermo Fisher ScientificSamlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Purity: 97%
CAS No.: 768-31-0
Molecular Formula: C6H10F3N
Formula Weight: 153.15
Flash Point: 28°(83°F)
Refractive Index: 1.3930
Odour: Amine-like
Boiling Point: 128-130°
UN No.: UN2733
MDL No.: MFCD02183561
Formel:
C₆H₁₀F₃N Kogepunkt: 128…130 °C Massefylde: 1,16 Flammepunkt: 28 °C (83 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD02183561 CAS-nummer: 768-31-0 ADR: 3,III |