4-Fluoropiperidine hydrochloride 95%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

H25755.03 H25755.MD
H25755.03EA 4090 DKK
H25755.03 H25755.MD
4-Fluoropiperidine hydrochloride 95%
4-Fluoropiperidine hydrochloride

Purity: 95% CAS No.: 57395-89-8 Molecular Formula: C5H10FN.HCl Formula Weight: 139.60 Melting Point: 161-163° MDL No.: MFCD03452786

Formel: C₅H₁₁ClFN
Molekylvægt: 139.6 g/mol
Smeltepunkt: 161…163 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD03452786
CAS-nummer: 57395-89-8

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR