Bis(2-mercaptoethyl)sulphide ≥90%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2,2'-Thiodiethanethiol, 2-Mercaptoethyl Sulphide

B22304.09 B22304.18 B22304.30
B22304.09EA 921 DKK
B22304.09 B22304.18 B22304.30
Bis(2-mercaptoethyl)sulphide ≥90%
Bis(2-mercaptoethyl)sulphide

Purity: 90+% CAS No.: 3570-55-6 Molecular Formula: HSCH2CH2SCH2CH2SH Formula Weight: 154.32 Flash Point: 90°(194°F) Boiling Point: 135-136°/10mm MDL No.: MFCD00004893

Formel: C₄H₁₀S₃
Molekylvægt: 154.30 g/mol
Kogepunkt: 135…136 °C (10 mmHg)
Massefylde: 1.183
Flammepunkt: 90 °C (194 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00004893
CAS-nummer: 3570-55-6
EINECS: 222-671-0

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR