2-Furylacetone 99%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

B22171.06 B22171.14
B22171.06EA 733 DKK
B22171.06 B22171.14
2-Furylacetone 99%
2-Furylacetone

Purity: 99% CAS No.: 6975-60-6 Molecular Formula: C7H8O2 Formula Weight: 124.14 Storage Temperature: 2°C to 8°C Flash Point: 65°(149°F) Odour: Stench Boiling Point: 179-180° MDL No.: MFCD02683084

Formel: C₇H₈O₂
Kogepunkt: 179…180 °C
Flammepunkt: 65 °C (149 °F)
MDL Number: MFCD02683084
CAS-nummer: 6975-60-6
EINECS: 230-234-0

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR