4-Methyl-5-vinylthiazole ≥98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

B21719.06 B21719.14 B21719.22
B21719.06EA 467 DKK
B21719.06 B21719.14 B21719.22
4-Methyl-5-vinylthiazole ≥98%
4-Methyl-5-vinylthiazole

Purity: 98+% CAS No.: 1759-28-0 Molecular Formula: C6H7NS Formula Weight: 125.19 Storage Temperature: 2°C to 8°C Flash Point: 70°(158°F) Refractive Index: 1.5680 Odour: Pleasant Melting Point: -15° Boiling Point: 67-68°/11mm Beilstein Reference: 107867 MDL No.: MFCD00005337

Formel: C₆H₇NS
Kogepunkt: 67…68 °C (11 mmHg)
Smeltepunkt: –15 °C
Massefylde: 1,095
Flammepunkt: 70 °C (158 °F)
MDL Number: MFCD00005337
CAS-nummer: 1759-28-0
EINECS: 217-160-4

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR