Ammonium iron(III) oxalate trihydrate 98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Iron ammonium oxalate, Ammonium ferric oxalate, Ammonium iron oxalate trihydrate

B21411.22 B21411.36
B21411.22EA 388 DKK
B21411.22 B21411.36
Ammonium iron(III) oxalate trihydrate 98%
Ammonium iron(III) oxalate trihydrate

Purity: 98% CAS No.: 13268-42-3 Molecular Formula: (NH4)3Fe(C2O4)3.3H2O Formula Weight: 428.07 (374.02anhy) UN No.: UN3288 MDL No.: MFCD00150137

Formel: (NH₄)₃Fe(C₂O₄)₃·3H₂O
Massefylde: 1,780
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00150137
CAS-nummer: 13268-42-3
EINECS: 238-090-0
UN nummer: 3288
ADR: 6.1,III
Merck Index: 14,517

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR