N-Ethylguanidinesulphate 98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 1-Ethylguanidine hemisulphate

B21315.06 B21315.14
B21315.06EA 614 DKK
B21315.06 B21315.14
N-Ethylguanidinesulphate 98%
N-Ethylguanidinesulphate

Purity: 98% CAS No.: 3482-86-8 Molecular Formula: [CH3CH2NHC(=NH)NH2]2.H2SO4 Formula Weight: 272.33 Odour: Odourless Melting Point: ca 245° dec. MDL No.: MFCD00013130

Formel: [CH₃CH₂NHC(=NH)NH₂]₂·H₂SO₄
Molekylvægt: 272.33 g/mol
Smeltepunkt: ∼ 245 °C (dec.)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00013130
CAS-nummer: 3482-86-8
EINECS: 222-467-1

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR