D-Tagatose 99%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
B21192.01
B21192.03
B21192.06
B21192.01EA
787
DKK
B21192.01
B21192.03
B21192.06
D-Tagatose 99%
D-Tagatose
Purity: 99%
CAS No.: 87-81-0
Molecular Formula: C6H12O6
Formula Weight: 180.16
Melting Point: 129-135°
Beilstein Reference: 1724555
Optical Rotation: -4.7° (c=1 in water)
MDL No.: MFCD00134449
Formel:
C₆H₁₂O₆ Smeltepunkt: 129…135 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00134449 CAS-nummer: 87-81-0 EINECS: 201-772-3 Merck Index: 14,9030 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...