Lactitol monohydrate 98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: (2S,3R,4R,5R)-4-(((2S,3R,4S,5R,6R)-3,4,5-Trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexane-1,2,3,5,6-pentaol hydrate

B21016.03 B21016.06
B21016.03EA 1160 DKK
B21016.03 B21016.06
Lactitol monohydrate 98%
Lactitol monohydrate

Purity: 98% CAS No.: 81025-04-9 Molecular Formula: C12H24O11.H2O Formula Weight: 362.33 (344.31anhy) Melting Point: 95-98° MDL No.: MFCD00150767

Formel: C₁₂H₂₄O₁₁·H₂O
Molekylvægt: 362.33 g/mol (344.31 anhy)
Smeltepunkt: 95…98 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00150767
CAS-nummer: 81025-04-9

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR